Merck
  • The effect of pH and ion channel modulators on human placental arteries.
The effect of pH and ion channel modulators on human placental arteries.
PloS one (2014-12-10)
Tayyba Y Ali, Fiona Broughton Pipkin, Raheela N Khan
ABSTRACT
Chorionic plate arteries (CPA) are located at the maternofetal interface where they are able to respond to local metabolic changes. Unlike many other types of vasculature, the placenta lacks nervous control and requires autoregulation for controlling blood flow. The placental circulation, which is of low-resistance, may become hypoxic easily leading to fetal acidosis and fetal distress however the role of the ion channels in these circumstances is not well-understood. Active potassium channel conductances that are subject to local physicochemical modulation may serve as pathways through which such signals are transduced. The aim of this study was to investigate the modulation of CPA by pH and the channels implicated in these responses using wire myography. CPA were isolated from healthy placentae and pre-contracted with U46619 before testing the effects of extracellular pH using 1 M lactic acid over the pH range 7.4-6.4 in the presence of a variety of ion channel modulators. A change from pH 7.4 to 7.2 produced a 29±3% (n = 9) relaxation of CPA which increased to 61±4% at the lowest pH of 6.4. In vessels isolated from placentae of women with pre-eclampsia (n = 6), pH responses were attenuated. L-methionine increased the relaxation to 67±7% (n = 6; p<0.001) at pH 6.4. Similarly the TASK 1/3 blocker zinc chloride (1 mM) gave a maximum relaxation of 72±5% (n = 8; p<0.01) which compared with the relaxation produced by the TREK-1 opener riluzole (75±5%; n = 6). Several other modulators induced no significant changes in vascular responses. Our study confirmed expression of several ion channel subtypes in CPA with our results indicating that extracellular pH within the physiological range has an important role in controlling vasodilatation in the human term placenta.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis