Merck
  • NAD attenuates oxidative DNA damages induced by amyloid beta-peptide in primary rat cortical neurons.
NAD attenuates oxidative DNA damages induced by amyloid beta-peptide in primary rat cortical neurons.
Free radical research (2014-04-01)
M-F Wu, J-H Yin, C-S Hwang, C-M Tang, D-I Yang
ABSTRACT
One major pathological hallmark of Alzheimer's disease (AD) is accumulation of senile plaques in patients' brains, mainly composed of amyloid beta-peptide (Aβ). Nicotinamide adenine dinucleotide (NAD) has emerged as a common mediator regulating energy metabolism, mitochondrial function, aging, and cell death, all of which are critically involved in neuronal demise observed in AD. In this work, we tested the hypothesis that NAD may attenuate Aβ-induced DNA damages, thereby conferring neuronal resistance to primary rat cortical cultures. We found that co-incubation of NAD dose-dependently attenuated neurotoxicity mediated by Aβ25-35 and Aβ1-42 in cultured rat cortical neurons, with the optimal protective dosage at 50 mM. NAD also abolished the formation of reactive oxygen species (ROS) induced by Aβ25-35. Furthermore, Aβs were capable of inducing oxidative DNA damages by increasing the extents of 8-hydroxy-2´-deoxyguanosine (8-OH-dG), numbers of apurinic/apyrimidinic (AP) sites, genomic DNA single-stranded breaks (SSBs), as well as DNA double-stranded breaks (DSBs)/fragmentation, which can all be attenuated upon co-incubation with NAD. Our results thus reveal a novel finding that NAD is protective against DNA damage induced by existing Aβ, leading ultimately to neuroprotection in primary cortical culture.
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)