Merck
  • Acetylsalicylic acid mitigates erythropoietin-associated blood pressure increase in nonuremic rats.
Acetylsalicylic acid mitigates erythropoietin-associated blood pressure increase in nonuremic rats.
Clinical and experimental hypertension (New York, N.Y. : 1993) (2014-10-15)
Maristela Böhlke, Rafael Giesteira, Claudia Castilho, Bartira Pinheiro, Maria Claudia Irigoyen, Carlos Eduardo Poli de Figueiredo
ABSTRACT
Approximately 30% of the chronic kidney disease patients using recombinant human erythropoietin (rhuEPO) have an increase in blood pressure (BP). Its mechanism and whether it depends on renal function remain unclear. There is early evidence that acetylsalicylic acid (ASA) prevents the rhuEPO-induced increase in BP. This study aims to verify whether very high doses of rhuEPO can increase BP in nonuremic rats and whether the co-administration of ASA can prevent it. Forty male Wistar rats were divided into four groups: placebo/placebo; placebo/rhuEPO 200 UI/kg thrice weekly; placebo/ASA 50 mg/kg daily; rhuEPO 200 UI/kg thrice weekly/ASA 50 mg/kg daily. Hematocrit was measured before and after and systolic BP was measured weekly by tail-cuff technique. Direct measurement of the BP was obtained at the end. The rhuEPO groups had higher final hematocrit (rhuEPO/placebo 56.7 ± 7.6, rhuEPO/ASA 56.7 ± 7.7; p < 0.001 versus placebo/placebo, 42.2 ± 4.7 and ASA/placebo 41.2 ± 4.2); and also increase in systolic BP (rhuEPO/placebo 135.1 ± 15.0, p = 0.01 and rhuEPO/ASA 127.2 ± 6.8, p = 0.02), whereas BP in rats from placebo/placebo (120.9 ± 5.0, p = 0.18) and placebo/ASA (124.6 ± 13.3, p = 0.12) groups remained unchanged. By direct measurement, the final BP was higher in rhuEPO/placebo (DBP 123.1 ± 12.0; SBP 157.4 ± 12.5; MBP 139.8 ± 11.9) than placebo/placebo (DBP 105.1 ± 11.5; SBP 141.0 ± 12.6; MBP 122.1 ± 12.1) and placebo/ASA groups (DBP 106.6 ± 8.1; SBP 141.5 ± 8.4, MBP 122.1 ± 7.2) (p < 0.05 by post hoc Bonferroni test ANOVA). The rhuEPO/ASA group (PAD 115.1 ± 11.4, PAS 147.4 ± 9.1, MBP 130.1 ± 10.3) was not different from other groups. The administration of very high doses of rhuEPO is associated with an increase in hematocrit and BP in nonuremic rats. The concomitant use of ASA mitigates the rhuEPO-associated BP increase.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)