Merck
  • Porcine synapsin 1: SYN1 gene analysis and functional characterization of the promoter.
Porcine synapsin 1: SYN1 gene analysis and functional characterization of the promoter.
FEBS open bio (2013-11-20)
Claus Hedegaard, Kasper Kjaer-Sorensen, Lone Bruhn Madsen, Carina Henriksen, Jamal Momeni, Christian Bendixen, Claus Oxvig, Knud Larsen
ABSTRACT
Synapsin 1 (SYN1) is a phosphoprotein involved in nerve signal transmission. The porcine SYN1 promoter orthologue was cloned and characterized to provide a means of expressing a transgene specifically in neurons. The nucleotide sequence of the promoter displayed a high degree of conservation of elements responsible for neuron-specific expression. Expression analysis of SYN1 demonstrated presence of transcript during embryonic development. Analysis of GFP expression in transgenic zebrafish embryos suggests that the pig SYN1 promoter directs expression in neuronal cells. Thus, the SYN1 promoter is a good candidate for use in the generation of pig models of human neurodegenerative disorders.
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
N-Phenylthiourea, ≥98%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.85%