Merck
Mutagenic potential of the isoflavone irilone in cultured V79 cells.
Toxicology letters (2015-02-24)
Anne Scheffler, Annette E Albrecht, Harald L Esch, Leane Lehmann
ABSTRACT
After consumption of red clover-based dietary supplements, plasma concentrations of the isoflavone irilone (IRI) equal that of the well-investigated daidzein. Since some isoflavones are genotoxic, the potential of IRI to induce mutations was investigated. Gene mutations were determined by hypoxanthine-guanine phosphoribosyltransferase (HPRT) assay and sequencing of mutant cDNA, chromosome and genome mutations by micronucleus assay complemented by immunochemical staining of centromere proteins and microtubules in cultured V79 cells. Cell proliferation was monitored by electronic cell counting, flow cytometry and fluorescence microscopy. IRI did not affect the mutant frequency in the Hprt locus but altered the mutation spectrum by increasing the proportion of deletions and decreasing that of base pair substitutions. Induction of chromosome mutations was supported by a slight but significant increase in the number of micronucleated cells containing chromosomal fragments despite activation of three cell cycle checkpoints possibly interfering with micronuclei formation. Moreover, IRI exhibited a strong aneugenic potential characterized by disrupted mitotic spindles, mitotic arrest, and asymmetrical cell divisions leading to chromosome loss, nuclear fragmentation as well as mitotic catastrophe. Thus, IRI might be another isoflavone to be taken into account in safety assessment of dietary supplements.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Magnesium methyl carbonate solution, 2.0 M in DMF
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Ethyl methanesulfonate, liquid
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
6-Thioguanine, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
4-Nitroquinoline N-oxide, ≥98%
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
6-Thioguanine, Hybri-Max, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma