Merck
  • Polymer enzyme conjugates as chiral ligands for sharpless dihydroxylation of alkenes in organic solvents.
Polymer enzyme conjugates as chiral ligands for sharpless dihydroxylation of alkenes in organic solvents.
Chembiochem : a European journal of chemical biology (2014-11-26)
Stefan Konieczny, Melanie Leurs, Joerg C Tiller
ABSTRACT
Conjugates of enzymes and poly(2-methyloxazoline) were used as organosoluble amphiphilic polymer nanocontainers for dissolving osmate, thereby converting the enzymes into organosoluble artificial metalloenzymes. These were shown to catalyze the dihydroxylation of different alkenes with high enantioselectivity. The highest selectivities, found for osmate complexed with laccase polymer-enzyme conjugates (PECs), even exceed those of classical Sharpless catalysts.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pyrogallol, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Methanol, analytical standard
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrocatechol, ≥99%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology