Merck
  • Histologic investigation of gingival epithelium implantation and the nonincision placement of miniscrews.
Histologic investigation of gingival epithelium implantation and the nonincision placement of miniscrews.
The International journal of oral & maxillofacial implants (2014-09-13)
Yang Zhang, Jieli Ni, Roger J Smales, Junqing Ma, Lin Wang
ABSTRACT
This study investigated whether the nonincision placement of miniscrews could lead to ectopic implantation of epithelium at the bone-implant interface and, if so, whether the epithelial cells could survive. The fate of grafted epithelial cells was also investigated. Ten healthy male beagle dogs were included in the study and were divided into three groups. Group 1 dogs (n = 3) were implanted with miniscrews without any other procedures. In group 2, epithelial cells were collected, infected by adeno-associated virus-2 containing green fluorescent protein (GFP), and injected into pilot holes drilled before miniscrew implantation in three dogs, and one dog was injected with epithelial cells as a negative control. In group 3, tiny gingival tissue pieces were injected into pilot holes drilled before miniscrew implantation. In group 1, hematoxylin-eosin and fluorescent immunohistochemical (IHC) staining of cytokeratin were performed on both undecalcified and decalcified slices. In groups 2 and 3, IHC staining of cytokeratin and apoptotic testing were performed. No ectopically implanted epithelial cells were found at the bone-implant interface of the miniscrews in group 1. In group 2, fluorescence emitted by cytokeratin antibody and by GFP was observed in specimens at 0 days, had decreased by 3 days, and had disappeared by 7 days. In group 3, areas of fluorescent cytokeratin-positive tissues were much smaller at 7 days than at 0 days, and none were found at 28 days. In groups 2 and 3, there were many apoptotic epithelial cells at the bone-implant interface at 3 days and 7 days, respectively. There was little possibility of implantation of epithelium into alveolar bone though placement of miniscrews with a nonincision procedure. Furthermore, even when epithelial cells or tissue was ectopically implanted, it was soon cleared through an apoptotic process.
Sigma-Aldrich
2-Ethoxyethyl acetate, 98%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20