Merck
  • Effects of HMG-CoA reductase inhibitors on the pharmacokinetics of nifedipine in rats: Possible role of P-gp and CYP3A4 inhibition by HMG-CoA reductase inhibitors.
Effects of HMG-CoA reductase inhibitors on the pharmacokinetics of nifedipine in rats: Possible role of P-gp and CYP3A4 inhibition by HMG-CoA reductase inhibitors.
Pharmacological reports : PR (2015-01-07)
Chong-Ki Lee, Jun-Shik Choi, Dong-Hyun Choi
ABSTRACT
This study aimed to investigate the effects of HMG-CoA reductase inhibitors on the pharmacokinetics of nifedipine in rats. We determined the pharmacokinetic parameters of nifedipine and dehydronifedipine in rats after oral and intravenous administration of nifedipine without and with HMG-CoA reductase inhibitors. We evaluated the effect of HMG-CoA reductase inhibitors on the activity of P-glycoprotein (P-gp) and cytochrome P450 (CYP)3A4. Atorvastatin, fluvastatin, pravastatin and simvastatin inhibited CYP3A4 activities; inhibitory concentration (IC50) values were 47.0, 5.2, 15.0 and 3.3 μM, respectively. Simvastatin and fluvastatin increased the cellular uptake of rhodamine-123. The area under the plasma concentration-time curve (AUC0-∞) and the peak plasma concentration (Cmax) of oral nifedipine were significantly increased by fluvastatin and simvastatin, respectively, compared to control group. The total body clearance (CL/F) of nifedipine after oral administration with fluvastatin and simvastatin were significantly decreased compared to those of control. The metabolite-parent AUC ratio (MR) of nifedipine with fluvastatin and simvastatin were significantly decreased, which suggested that fluvastatin and simvastatin inhibited metabolism of nifedipine, respectively. The AUC0-∞ of intravenouse nifedipine with fluvastatin and simvastatin was significantly higher than that of the control group. The increased bioavailability of nifedipine may be mainly due to inhibition of both P-gp in the small intestine and CYP3A subfamily-mediated metabolism of nifedipine in the small intestine and/or in the liver and to the reduction of the CL/F of nifedipine by fluvastatin and simvastatin.
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
2,2,4-Trimethylpentane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
Supelco
Simvastatin, analytical standard
Supelco
2,2,4-Trimethylpentane, analytical standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2,2,4-Trimethylpentane, ACS reagent, ≥99.0%
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Triethylamine, ≥99%