Merck
  • The application of metabolite profiling to Mycobacterium spp.: determination of metabolite changes associated with growth.
The application of metabolite profiling to Mycobacterium spp.: determination of metabolite changes associated with growth.
Journal of microbiological methods (2014-08-12)
Margit Drapal, Laura Perez-Fons, Paul R Wheeler, Paul D Fraser
ABSTRACT
In order to decipher the complex biological networks underlying biochemical and physiological processes, cellular regulation at all levels must be studied. The metabolites determined by metabolomics represent the end-point of cellular regulation and thus vital components of any integrative network. In the case of pathogenic agents such as Mycobacterium tuberculosis metabolomics offers an ideal opportunity to gain a better understanding of how this species adapts to environmental conditions and antimicrobial treatments. In the present study a metabolite profiling protocol for Mycobacterium including optimised quenching, extraction and analysis has been devised. These methods have been applied to three different Mycobacterium spp. demonstrating potential translation across the genus. Steady-state levels of metabolites during growth have been determined for Mycobacterium smegmatis, Mycobacterium phlei and Mycobacterium bovis BCG (Bacillus Calmette-Guérin). The changes of designated biomarkers emphasised phenotypical differences (e.g. nitrogen metabolism) and similarities (e.g. cysteine biosynthesis) between the bacteria. Each time point showed distinguishable metabolic characteristics from early lag to late stationary phase/beginning of non-replicating phase. The combination of the metabolic results with published "omics" data indicated that transcription appeared to be the most predominant mode of cellular regulation utilised by these bacteria studied.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
N-Methyl-bis(trifluoroacetamide), derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
N-Methyl-bis(trifluoroacetamide), derivatization grade (GC derivatization), LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Trifluoroacetamide, 97%
Supelco
Acetic anhydride, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%