Merck
  • Gene expression profile analysis of human mesenchymal stem cells from herniated and degenerated intervertebral discs reveals different expression of osteopontin.
Gene expression profile analysis of human mesenchymal stem cells from herniated and degenerated intervertebral discs reveals different expression of osteopontin.
Stem cells and development (2014-09-10)
Giovanni Marfia, Stefania Elena Navone, Clara Di Vito, Silvia Tabano, Lorenzo Giammattei, Andrea Di Cristofori, Roberta Gualtierotti, Carlo Tremolada, Mario Zavanone, Manuela Caroli, Francesco Torchia, Monica Miozzo, Paolo Rampini, Laura Riboni, Rolando Campanella
ABSTRACT
Gene expression analysis provides an effective methodology to identify clinically relevant genes implicated in intervertebral disc (IVD) pathology. The analysis of gene profile in mesenchymal stem cells (MSCs) from human herniated IVD (H-IVD) and degenerated IVD (D-IVD) has not yet been investigated. We present in this study a characterization of MSCs isolated from clinically categorized H-IVD and D-IVD disc samples. H-IVD-MSCs and D-IVD-MSCs showed multipotent mesenchymal differentiation ability, expressing positivity for adipogenic, osteogenic, and chondrogenic markers with an immunophenotypical profile representative of MSCs. FACS analyses revealed a higher expression of CD44 in D-IVD-MSCs compared to H-IVD-MSCs. Gene expression profile revealed that most genes under investigation displayed large variations and were not significantly different in the two types of analyzed IVD-MSCs. Conversely, the gene expression of osteopontin (OPN), a protein involved in bone matrix mineralization and extracellular matrix destruction, was found markedly increased (more than 400-fold) in D-IVD-MSCs compared to H-IVD-MSCs. Moreover, the OPN protein expression was detectable only in D-IVD-MSCs, and its levels were directly related with D-IVD severity. These findings suggest that an abnormal expression of OPN in D-IVD-MSCs occurs and plays a pivotal role in the pathophysiological process of human disc degeneration. We speculate that the regulation of the OPN pathway might be a therapeutic target to counteract disc degeneration.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.