Merck
  • An UPLC-MS/MS method for quantitative profiling of bile acids in sea lamprey plasma and tissues.
An UPLC-MS/MS method for quantitative profiling of bile acids in sea lamprey plasma and tissues.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-16)
Huiyong Wang, Chu-Yin Yeh, Ke Li, Yu-Wen Chung-Davidson, Weiming Li
ABSTRACT
Bile acids (BAs) have recently gained more attention because of their diverse roles from digestion to signaling. Simultaneous analyses of various BAs in biological samples are challenging due to their structural similarity, relatively low concentrations, and the presence of isomeric forms. In this study, we report a simple and sensitive UPLC-MS/MS method for simultaneous quantifications of 13 BAs including four unique sea lamprey BAs in sea lamprey plasma, liver, intestine, and gills. A straightforward protein precipitation (PPT) method was used to extract BAs from the biological samples. Separation of all target analytes was achieved on a reverse-phase UPLC column in 15min, and detection was carried out on MS/MS with ESI in the negative ionization mode. This method was validated regarding its linearity, limits of detection (LOD), recovery, matrix effect, reproducibility, accuracy and precision. Significant improvements compared to previous LC-MS/MS methods were observed as a result of the application of UPLC and extensive optimization of experimental conditions. The method showed excellent linearity with high regression coefficients (>0.99) over a range of 0.5-1000ng/mL and LODs ranged from 0.009 to 0.11ng/mL. The applications of the developed method demonstrated that it simultaneously determined all target BAs in different biological sample matrices with excellent sensitivity, selectivity and reproducibility.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis