Merck
  • Myotubes from lean and severely obese subjects with and without type 2 diabetes respond differently to an in vitro model of exercise.
Myotubes from lean and severely obese subjects with and without type 2 diabetes respond differently to an in vitro model of exercise.
American journal of physiology. Cell physiology (2015-01-23)
Yuan Z Feng, Nataša Nikolić, Siril S Bakke, Eili T Kase, Kari Guderud, Jøran Hjelmesæth, Vigdis Aas, Arild C Rustan, G Hege Thoresen
ABSTRACT
Exercise improves insulin sensitivity and oxidative capacity in skeletal muscles. However, the effect of exercise on substrate oxidation is less clear in obese and type 2 diabetic subjects than in lean subjects. We investigated glucose and lipid metabolism and gene expression after 48 h with low-frequency electrical pulse stimulation (EPS), as an in vitro model of exercise, in cultured myotubes established from lean nondiabetic subjects and severely obese subjects (BMI ≥ 40 kg/m(2)) with and without type 2 diabetes. EPS induced an increase in insulin sensitivity but did not improve lipid oxidation in myotubes from severely obese subjects. Thus, EPS-induced increases in insulin sensitivity and lipid oxidation were positively and negatively correlated to BMI of the subjects, respectively. EPS enhanced oxidative capacity of glucose in myotubes from all subjects. Furthermore, EPS reduced mRNA expression of slow fiber-type marker (MYH7) in myotubes from diabetic subjects; however, the protein expression of this marker was not significantly affected by EPS in either of the donor groups. On the contrary, mRNA levels of interleukin-6 (IL-6) and IL-8 were unaffected by EPS in myotubes from diabetic subjects, while IL-6 mRNA expression was increased in myotubes from nondiabetic subjects. EPS-stimulated mRNA expression levels of MYH7, IL-6, and IL-8 correlated negatively with subjects' HbA1c and/or fasting plasma glucose, suggesting an effect linked to the diabetic phenotype. Taken together, these data show that myotubes from different donor groups respond differently to EPS, suggesting that this effect may reflect the in vivo characteristics of the donor groups.
Product Description
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method