Merck
  • Simultaneous quantification of IMPDH activity and purine bases in lymphocytes using LC-MS/MS: assessment of biomarker responses to mycophenolic acid.
Simultaneous quantification of IMPDH activity and purine bases in lymphocytes using LC-MS/MS: assessment of biomarker responses to mycophenolic acid.
Therapeutic drug monitoring (2013-09-26)
Nils Tore Vethe, Ali Mohamad Ali, Pål Aarstrand Reine, Anders M Andersen, Sara Bremer, Pål-Dag Line, Helge Rootwelt, Stein Bergan
ABSTRACT
The development of biomarkers describing the individual responses to the immunosuppressant mycophenolic acid (MPA) has focused on the target enzyme activity [inosine 5'-monophosphate dehydrogenase (IMPDH)]. An extended strategy is to quantify the metabolic consequences of IMPDH inhibition. The aim of this study was to develop an assay for quantification of IMPDH activity and related purine bases and to provide preliminary data on the behavior of these biomarkers during clinical exposure to MPA. Liquid chromatography-mass spectrometry was used to determine xanthine (IMPDH activity in incubated cell lysate), hypoxanthine, guanine, and adenine derived from free nucleotides in lymphocytes. Analytical performance was assessed, and the biomarkers were examined in CD4⁺ cells from 2 groups: Healthy individuals in a single-dose MPA study (n = 5) and liver transplant recipients on MPA therapy (n = 15). Coefficients of variation between series were below 10% and 15% for measurement of the purines and IMPDH activity, respectively. Although IMPDH was inhibited, the purine levels increased in response to MPA in 3 of the 5 healthy individuals, and this positive response seemed to be associated with IMPDH1 c.579 + 119 G/G and c.580 - 106 G/G. In the liver transplant study, guanine was not reduced in response to the transient drop in IMPDH activity after MPA dosing. However, there were trends toward decrease in guanine and elevation of hypoxanthine during prolonged MPA therapy. The guanine/hypoxanthine ratio (median) was 37% lower and the adenine level was 21% lower at day 17 compared with day 4 after transplantation. The assay allows precise quantification of IMPDH activity, hypoxanthine, guanine, and adenine in lymphocytes. Some individuals may possess a counteracting purine response to the MPA-mediated inhibition of IMPDH. Reduction of the guanine/hypoxanthine ratio may be related to prolonged inhibition of IMPDH and seems as an intriguing pharmacodynamic biomarker for MPA.
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium acetate solution, BioUltra, Molecular Biology, 5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Adenine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium acetate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Potassium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Potassium acetate, 98%
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard