Merck
  • Multi-modal transfection agent based on monodisperse magnetic nanoparticles for stem cell gene delivery and tracking.
Multi-modal transfection agent based on monodisperse magnetic nanoparticles for stem cell gene delivery and tracking.
Biomaterials (2014-06-02)
Wooram Park, Han Na Yang, Daishun Ling, Hyeona Yim, Kyoung Sub Kim, Taeghwan Hyeon, Kun Na, Keun-Hong Park
ABSTRACT
Directing the controlled differentiation and tracking of stem cells is essential to achieve successful stem cell therapy. In this work, we describe a multi-modal (MR/optical) transfection agent (MTA) for efficient gene delivery and cell tracking of human mesenchymal stem cells (hMSCs). The MTA was synthesized through a facile two-step approach with 1) ligand exchange of a catechol-functionalized polypeptide (CFP) and 2) chemical immobilization of fluorescence labelled cationic polymer via aminolysis reaction. Cationic polymer-immobilized MTAs with size of ~40 nm exhibit greatly enhanced colloidal stability in aqueous solution. In addition, the MTAs were capable of binding DNA molecules for transfection. The MTA/pDNA complex showed relatively good transfection efficiency in hMSCs (compared to the commercial transfection agent, Lipofectamine) and good biocompatibility. MTA-treated hMSCs were successfully visualized after transplantation via MR and optical imaging system over 14 days. These studies highlight the challenges associated with the potential advantages of designing multi-modal nanostructured materials as tools for genetic materials delivery and cell-tracking in stem cell therapy.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)
Supelco
Chloroform, analytical standard
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, natural, 99%, FG