Merck
  • Expression of the p53 target Wig-1 is associated with HPV status and patient survival in cervical carcinoma.
Expression of the p53 target Wig-1 is associated with HPV status and patient survival in cervical carcinoma.
PloS one (2014-11-08)
Li-Di Xu, Susanne Muller, Srinivasan R Thoppe, Fredrik Hellborg, Lena Kanter, Mikael Lerner, Biying Zheng, Svetlana Bajalica Lagercrantz, Dan Grandér, Keng Ling Wallin, Klas G Wiman, Catharina Larsson, Sonia Andersson
ABSTRACT
The p53 target gene WIG-1 (ZMAT3) is located in chromosomal region 3q26, that is frequently amplified in human tumors, including cervical cancer. We have examined the status of WIG-1 and the encoded Wig-1 protein in cervical carcinoma cell lines and tumor tissue samples. Our analysis of eight cervical cancer lines (Ca Ski, ME-180, MS751, SiHa, SW756, C-4I, C-33A, and HT-3) by spectral karyotype, comparative genomic hybridization and Southern blotting revealed WIG-1 is not the primary target for chromosome 3 gains. However, WIG-1/Wig-1 were readily expressed and WIG-1 mRNA expression was higher in the two HPV-negative cervical cell lines (C33-A, HT-3) than in HPV-positive lines. We then assessed Wig-1 expression by immunohistochemistry in 38 cervical tumor samples. We found higher nuclear Wig-1 expression levels in HPV-negative compared to HPV positive cases (p = 0.002) and in adenocarcinomas as compared to squamous cell lesions (p<0.0001). Cases with moderate nuclear Wig-1 staining and positive cytoplasmic Wig-1 staining showed longer survival than patients with strong nuclear and negative cytoplasmic staining (p = 0.042). Nuclear Wig-1 expression levels were positively associated with age at diagnosis (p = 0.023) and histologic grade (p = 0.034). These results are consistent with a growth-promoting and/or anti-cell death function of nuclear Wig-1 and suggest that Wig-1 expression can serve as a prognostic marker in cervical carcinoma.
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.