Merck
  • Laboratory measurements of the oral direct factor Xa inhibitor edoxaban: comparison of prothrombin time, activated partial thromboplastin time, and thrombin generation assay.
Laboratory measurements of the oral direct factor Xa inhibitor edoxaban: comparison of prothrombin time, activated partial thromboplastin time, and thrombin generation assay.
American journal of clinical pathology (2015-01-18)
Yoshiyuki Morishima, Chikako Kamisato
ABSTRACT
Edoxaban, an oral direct factor Xa inhibitor, does not require routine monitoring. However, assessment of the anticoagulant effects may be required in certain situations. We investigated the effects of edoxaban on prothrombin time (PT), activated partial thromboplastin time (aPTT), and thrombin generation using human platelet-poor plasma (PPP) or platelet-rich plasma (PRP). Edoxaban concentration-dependently prolonged PT and aPTT. There was a considerable variation in the magnitude of PT prolongation among the reagents used. The variability in aPTT prolongation among the reagents was smaller than that of PT. Edoxaban concentration-dependently inhibited thrombin generation, with a more potent effect seen in PPP than in PRP. Thrombin generation assay was three times more sensitive to edoxaban than PT and aPTT. PT had disadvantages of a large variability among different PT reagents. aPTT could be used as a conventional and convenient test with a smaller variation among reagents. Thrombin generation was the most sensitive assay.
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Supelco
Ellagic acid, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture