Merck
  • Epithelial-derived nuclear IL-33 aggravates inflammation in the pathogenesis of reflux esophagitis.
Epithelial-derived nuclear IL-33 aggravates inflammation in the pathogenesis of reflux esophagitis.
Journal of gastroenterology (2014-08-19)
Jing Shan, Tadayuki Oshima, Taichiro Muto, Koubun Yasuda, Hirokazu Fukui, Jiro Watari, Kenji Nakanishi, Hiroto Miwa
ABSTRACT
IL-33 is a new tissue-derived cytokine constitutively expressed in epithelial cells and plays a role in sensing damage caused by inflammatory diseases. The function of IL-33 in the esophageal mucosa has not been previously described. Accordingly, we examined the expression of IL-33 and its role in the pathogenesis of reflux esophagitis (RE). IL-33 in the esophageal mucosa of RE patients and in an in vitro stratified normal esophageal squamous epithelial model was examined at the messenger RNA and protein levels. The correlation of the level of IL-33 and IL-8 or IL-6 was examined. Cell layers were stimulated with bile acids and cytokines. IL-33 was knocked down by small interfering RNA (siRNA). Pharmacological inhibitors and signal transducer and activator of transcription 1 (STAT1) siRNA were used. IL-33 was significantly upregulated in RE patients, and was located in the nuclei of basal and suprabasal layers. Upregulated IL-33 messenger RNA expression was correlated with IL-8 and IL-6 expression. In vitro, IL-33 was upregulated in the nuclei of basal and suprabasal layers by interferon-γ (IFNγ), and the upregulation was aggravated by the combination of deoxycholic acid (DCA) and IFNγ. IL-33 knockdown dampened IFNγ- and DCA-induced IL-8 and IL-6 production. IFNγ-induced IL-33 was inhibited by a Janus kinase inhibitor, a p38 mitogen-activated protein kinase inhibitor, and STAT1 siRNA. Nuclear IL-33 is upregulated in erosive mucosa of RE patients and is correlated with IL-8 and IL-6 levels. The normal esophageal epithelial model enables us to show for the first time that epithelial-cell-derived nuclear but not exogenous IL-33 is located upstream of the production of inflammatory cytokines and can aggravate the inflammation.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Chenodeoxycholic acid
Chenodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
Epigallocatechin gallate, primary reference standard
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(−)-Epigallocatechin gallate, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)