Merck
  • Ameliorative effect of propolis against cyclophosphamide-induced toxicity in mice.
Ameliorative effect of propolis against cyclophosphamide-induced toxicity in mice.
Pharmaceutical biology (2014-10-08)
Sabry A El-Naggar, Abeer A Alm-Eldeen, Mousa O Germoush, Kamal F El-Boray, Hassan A Elgebaly
ABSTRACT
Cyclophosphamide (CTX) is a common anticancer agent used for the treatment of several malignancies. However, upon treatment, it induces severe toxicity due to its oxidative stress capability. Propolis, a natural product collected by honey bees, has shown several biological activities, such as free radical scavenging and antioxidant agent. This study elucidates the protective effects of propolis against CTX-induced changes in mice. Forty-eight male Swiss albino mice were divided into four groups; group 1 was intraperitoneally (i.p.) injected with 200 µL of phosphate buffer saline (PBS), group 2 was injected with 100 mg/kg/d propolis, group 3 was injected with a single dose of CTX (200 mg/kg), and group 4 was injected with a single dose of CTX (200 mg/kg) followed by propolis (100 mg/kg) for 7 consecutive days. After 12 d, mice were bled and then sacrificed to analyze the hematological, biochemical, and histological parameters. The results indicated that CTX-injected mice showed an increase in the levels of alanine aminotransferase (ALT), aspartate aminotransferase (AST), urea, and creatinine and a decrease in the total number of white blood cells (WBCs) and platelets. Moreover, dramatically changes in the histological architectures of the liver and kidney were observed. The mice that were injected with CTX/propolis showed an improvement in the levels of ALT, AST, urea, creatinine, WBCs, and platelets. Moreover, the histological picture of the liver and kidney was significantly improved. In conclusion, propolis might be considered an effective agent in ameliorating the toxicity resulted from CTX treatment.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)