Merck
  • D-4F, an apolipoprotein A-I mimetic peptide, protects human umbilical vein endothelial cells from oxidized low-density lipoprotein-induced injury by preventing the downregulation of pigment epithelium-derived factor expression.
D-4F, an apolipoprotein A-I mimetic peptide, protects human umbilical vein endothelial cells from oxidized low-density lipoprotein-induced injury by preventing the downregulation of pigment epithelium-derived factor expression.
Journal of cardiovascular pharmacology (2014-04-09)
Jie Liu, Shutong Yao, Shuxia Wang, Peng Jiao, Guohua Song, Yang Yu, Ping Zhu, Shucun Qin
ABSTRACT
To investigate the protective effects of D-4F, an apolipoprotein A-I mimetic peptide, on oxidized low-density lipoprotein (ox-LDL)-induced injury of vascular endothelial cells and the potential role of pigment epithelium-derived factor (PEDF). Cytotoxicity was assessed by the apoptotic rate, 3-(4,5-dimethylthiazol-2-y-l)-2,5-diphenyl-2H-tetrazolium bromide assay, and lactate dehydrogenase release. PEDF levels were analyzed with Western blot and quantitative real-time polymerase chain reaction. Redox status was measured by the levels of the reactive oxygen species, malondialdehyde, superoxide dismutase, and nitric oxide. Ox-LDL reduced cell viability and induced apoptosis and LDH release from human umbilical vein endothelial cells, but the cytotoxic effects of ox-LDL were significantly inhibited by pretreatment with D-4F. Additionally, D-4F could scavenge intracellular reactive oxygen species, suppress the production of lipid peroxides, and improve endogenous antioxidant activity. Ox-LDL decreased PEDF expression in human umbilical vein endothelial cells in a concentration-dependent manner, and this decrease was markedly attenuated by D-4F. However, silencing PEDF by short interfering RNA blocked the inhibitory effects of D-4F on ox-LDL-induced oxidative stress and cellular injury. D-4F effectively protects vascular endothelial cells against ox-LDL-induced injury by preventing the downregulation of PEDF expression.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
MISSION® esiRNA, targeting human SERPINF1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Serpinf1
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%