Merck
  • Neochromine S5 improves contact hypersensitivity through a selective effect on activated T lymphocytes.
Neochromine S5 improves contact hypersensitivity through a selective effect on activated T lymphocytes.
Biochemical pharmacology (2014-09-15)
Zhe Gao, Yuxiang Ma, Dan Zhao, Xiong Zhang, Hang Zhou, Hailiang Liu, Yang Zhou, Xuefeng Wu, Yan Shen, Yang Sun, Jianxin Li, Xudong Wu, Qiang Xu
ABSTRACT
Strategy on activated T cells is an effective treatment for T cell mediated diseases. By using a synthesized chromone derivative, we examined its effects on the activated T cells. This compound, (Z)-1,3-dihydroxy-9-methyl-13H-benzo[b]chromeno[3,2-f][1,4]oxazepin-13-one (neochromine S5), exhibited immunosuppressive activity in vitro and in vivo. Interestingly, neochromine S5 selectively inhibited proliferation and induced apoptosis in T lymphocytes activated by concanavalin A (Con A) in a dose-dependent manner but not in naïve T lymphocytes, distinct from quercetin. This compound triggered mitochondrial apoptotic pathway including cleavage of caspase 3, caspase 9 and PARP, downregulation of bcl-2 and release of cytochrome c in activated T cells, but did not affect ER stress or Fas signals. In addition, neochromine S5 downregulated the expression of CD25 and CD69 and the production of inflammatory cytokines, including TNFα, IFNγ and IL-2, improved ear swelling in mice with contact hypersensitivity, reduced CD4(+) T cells infiltration, and increased apoptosis of isolated T lymphocytes from peripheral lymph nodes. Moreover, neochromine S5 showed no effect on the weight of mice and their immune organs, while dexamethasone caused a significant weight loss. Taken together, our results suggest that neochromine S5 exerts a unique anti-inflammatory activity mainly through a selective effect on activated T cells, which is different from the current immunosuppressant, dexamethasone.
Product Description
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Gly-Gly, ≥99% (titration)
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
Gly-Gly, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Gly-Gly, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Gly-Gly, BioPerformance Certified, suitable for cell culture, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Chloro-1,3,5-trinitrobenzene, ≥98.0% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Picrylsulfonic acid solution, 1 M in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)