Merck
  • Decidual natural killer cells regulate vessel stability: implications for impaired spiral artery remodelling.
Decidual natural killer cells regulate vessel stability: implications for impaired spiral artery remodelling.
Journal of reproductive immunology (2015-05-26)
Rupsha Fraser, Guy St J Whitley, Baskaran Thilaganathan, Judith E Cartwright
ABSTRACT
Decidual NK (dNK) cells are present during uterine spiral artery remodelling, an event that is crucial for successful placentation and the provision of an adequate blood supply to the developing fetus. Spiral artery remodelling is impaired in the pregnancy complication pre-eclampsia. Although dNK cells are known to play active roles at the maternal-fetal interface, little is known about their effect on endothelial integrity, an important component of vessel stability. We present a study in which we have modelled dNK-endothelium interactions, using first-trimester dNK cells isolated from both normal pregnancies and those with impaired spiral artery remodelling. dNK cells were isolated from first-trimester pregnancies, screened by uterine artery Doppler ultrasound to determine resistance indices (RI) that relate to the extent of spiral artery remodelling. dNK culture supernatant from normal-RI pregnancies (but not high-RI pregnancies) destabilised endothelial tube-like structures in Matrigel, and normal-RI dNK cells induced endothelial intercellular adhesion molecule-1 and tumour necrosis factor-α expression to a greater extent than high-RI dNK cells. We have established a functional role for dNK cells in the disruption of endothelial structures and have suggested how impairment of this process may be contributing to the reduced vessel remodelling in pregnancies with a high uterine artery resistance index. These findings have implications for our understanding of the pathology of pre-eclampsia and other pregnancy disorders where remodelling is impaired.
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma