Merck
  • Chondroitin sulfate proteoglycans impede myelination by oligodendrocytes after perinatal white matter injury.
Chondroitin sulfate proteoglycans impede myelination by oligodendrocytes after perinatal white matter injury.
Experimental neurology (2015-04-12)
Ying-Ping Deng, Yi Sun, Lan Hu, Zhi-Hua Li, Quan-Mei Xu, Yi-Ling Pei, Zhi-Heng Huang, Zhen-Gang Yang, Chao Chen
ABSTRACT
Hypomyelination is the major cause of neurodevelopmental deficits that are associated with perinatal white matter injury. Chondroitin sulfate proteoglycans (CSPGs) are known to exert inhibitory effects on the migration and differentiation of oligodendrocytes (OLs). However, few studies describe the roles of CSPGs in myelination by OLs and the cognitive dysfunction that follows perinatal white matter injury. Here, we examined the alterations in the expression of CSPGs and their functional impact on the maturation of OLs and myelination in a neonatal rat model of hypoxic-ischemic (HI) brain injury. Three-day-old Sprague-Dawley rats underwent a right common carotid artery ligation and were exposed to hypoxia (6% oxygen for 2.5h). Rats were given chondroitinase ABC (cABC) via an intracerebroventricular injection to digest CSPGs. Animals were sacrificed at 7, 14, 28 and 56days after HI injury and the accompanying surgical procedure. We found that the expression of CSPGs was significantly up-regulated in the cortical regions surrounding the white matter after HI injury. cABC successfully degraded CSPGs in the rats that received cABC. Immunostaining showed decreased expression of the pre-oligodendrocyte marker O4 in the cingulum, external capsule and corpus callosum in HI+cABC rats compared to HI rats. However HI+cABC rats exhibited greater maturation of OLs than did HI rats, with increased expression of O1 and myelin basic protein in the white matter. Furthermore, using electron microscopy, we demonstrated that myelin formation was enhanced in HI+cABC rats, which had an increased number of myelinated axons and decreased G-ratios of myelin compared to HI rats. Finally, HI+cABC rats performed better in the Morris water maze task than HI rats, which indicates an improvement in cognitive ability. Our results suggest that CSPGs inhibit both the maturation of OLs and the process of myelination after neonatal HI brain injury. The data also raise the possibility that modifying CSPGs may repair this type of lesion associated with demyelination.
Product Description
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hydriodic acid, distilled, 57 wt. %, 99.999% trace metals basis
Sigma-Aldrich
Ethylene glycol 5 M solution