Merck
  • Comparison of sulfo-conjugated and gluco-conjugated urinary metabolites for detection of methenolone misuse in doping control by LC-HRMS, GC-MS and GC-HRMS.
Comparison of sulfo-conjugated and gluco-conjugated urinary metabolites for detection of methenolone misuse in doping control by LC-HRMS, GC-MS and GC-HRMS.
Journal of mass spectrometry : JMS (2015-08-12)
A G Fragkaki, Y S Angelis, P Kiousi, C G Georgakopoulos, E Lyris
ABSTRACT
Methenolone (17β-hydroxy-1-methyl-5α-androst-1-en-3-one) misuse in doping control is commonly detected by monitoring the parent molecule and its metabolite (1-methylene-5α-androstan-3α-ol-17-one) excreted conjugated with glucuronic acid using gas chromatography-mass spectrometry (GC-MS) and liquid chromatography mass spectrometry (LC-MS) for the parent molecule, after hydrolysis with β-glucuronidase. The aim of the present study was the evaluation of the sulfate fraction of methenolone metabolism by LC-high resolution (HR)MS and the estimation of the long-term detectability of its sulfate metabolites analyzed by liquid chromatography tandem mass spectrometry (LC-HRMSMS) compared with the current practice for the detection of methenolone misuse used by the anti-doping laboratories. Methenolone was administered to two healthy male volunteers, and urine samples were collected up to 12 and 26 days, respectively. Ethyl acetate extraction at weak alkaline pH was performed and then the sulfate conjugates were analyzed by LC-HRMS using electrospray ionization in negative mode searching for [M-H](-) ions corresponding to potential sulfate structures (comprising structure alterations such as hydroxylations, oxidations, reductions and combinations of them). Eight sulfate metabolites were finally detected, but four of them were considered important as the most abundant and long term detectable. LC clean up followed by solvolysis and GC/MS analysis of trimethylsilylated (TMS) derivatives reveal that the sulfate analogs of methenolone as well as of 1-methylene-5α-androstan-3α-ol-17-one, 3z-hydroxy-1β-methyl-5α-androstan-17-one and 16β-hydroxy-1-methyl-5α-androst-1-ene-3,17-dione were the major metabolites in the sulfate fraction. The results of the present study also document for the first time the methenolone sulfate as well as the 3z-hydroxy-1β-methyl-5α-androstan-17-one sulfate as metabolites of methenolone in human urine. The time window for the detectability of methenolone sulfate metabolites by LC-HRMS is comparable with that of their hydrolyzed glucuronide analogs analyzed by GC-MS. The results of the study demonstrate the importance of sulfation as a phase II metabolic pathway for methenolone metabolism, proposing four metabolites as significant components of the sulfate fraction.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Ammonium iodide, 99.999% trace metals basis
Sigma-Aldrich
1,4-Dithioerythritol, BioXtra, ≥99.0%
Sigma-Aldrich
Heptacosafluorotributylamine, liquid
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, Molecular Biology, ≥99.0%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
1,4-Dithioerythritol, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%