Merck
  • Evaluation of two molecularly imprinted polymers for the solid-phase extraction of natural, synthetic and mycoestrogens from environmental water samples before liquid chromatography with mass spectrometry.
Evaluation of two molecularly imprinted polymers for the solid-phase extraction of natural, synthetic and mycoestrogens from environmental water samples before liquid chromatography with mass spectrometry.
Journal of separation science (2015-05-28)
Javier González-Sálamo, Bárbara Socas-Rodríguez, Javier Hernández-Borges, María Del Mar Afonso, Miguel Ángel Rodríguez-Delgado
ABSTRACT
In this work, we have compared the selectivity of two commercial molecularly imprinted polymers (AFFINIMIP®SPE Estrogens and AFFINIMIP®SPE Zearalenone) for the extraction of 12 estrogenic compounds of interest (i.e. 17α-estradiol, 17β-estradiol, estrone, hexestrol, 17α-ethynylestradiol, diethylstibestrol, dienestrol, zearalenone, α-zearalanol, β-zearalanol, α-zearalenol and β-zearalenol) from different water samples. High-performance liquid chromatography coupled with ion trap mass spectrometry with electrospray ionization was used for their determination. Results showed that although both molecularly imprinted polymeric cartridges were specifically designed for different groups of analytes (natural estrogens like estradiol in the first case and zearalenone derivatives in the second) they nearly have the same extraction performance (with recovery values in the range 65-101%) for the same analytes in Milli-Q water because of the cross-reactivity of the polymer. However, when more complex water samples were analyzed, it was clear that the behavior was different and that the AFFINIMIP®SPE Estrogens showed less cross-reactivity than the other cartridge. Validation of the proposed methodology with both cartridges revealed that the extraction was reproducible and that the final limits of detection of the proposed method were in the low ng/L range.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Estrone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hexarelin, ≥90% (HPLC)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%