Merck
  • Serotonin, but not dopamine, controls the stress response and anxiety-like behavior in the crayfish Procambarus clarkii.
Serotonin, but not dopamine, controls the stress response and anxiety-like behavior in the crayfish Procambarus clarkii.
The Journal of experimental biology (2015-07-04)
Pascal Fossat, Julien Bacqué-Cazenave, Philippe De Deurwaerdère, Daniel Cattaert, Jean-Paul Delbecque
ABSTRACT
In the animal kingdom, biogenic amines are widespread modulators of the nervous system that frequently interact to control mood. Our previous investigations in crayfish (Procambarus clarkii) have established that stress induces changes in brain serotonin (5-HT) concentrations that are responsible for the appearance of anxiety-like behavior (ALB). Here, we further analyze the roles of 5-HT and another biogenic amine, dopamine (DA), on the crayfish response to stress. We show that the intensity of crayfish ALB depends on the intensity of stressful stimulation and is associated with increased concentrations of 5-HT in the brain. These 5-HT levels were significantly correlated, before, as well as after stress, with those of DA, which were approximately 3- to 5-times less abundant. However, whereas the degree of ALB was clearly correlated with brain 5-HT concentrations, it was not significantly correlated with DA. Moreover, in contrast to injections of 5-HT, DA injections were not able to elicit a stress response or ALB. In addition, 5-HT and DA levels were not modified by treatment with the anxiolytic chlordiazepoxide, confirming that suppression of ALB by this GABA-A receptor ligand acts downstream and is independent of changes in crayfish bioamine levels. Our study also provides evidence that the anxiogenic effect of 5-HT injections can be prevented by a preliminary injection of 5-HT antagonists. Altogether, our results emphasize that the rises in brain concentrations of 5-HT, but not DA, play a role in controlling the induction and the intensity of crayfish ALB.
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Mianserin hydrochloride
Sigma-Aldrich
Methysergide maleate salt, solid
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology