Merck
  • Stages of Cell Cannibalism--Entosis--in Normal Human Keratinocyte Culture.
Stages of Cell Cannibalism--Entosis--in Normal Human Keratinocyte Culture.
Biochemistry. Biokhimiia (2015-11-30)
A S Garanina, L A Khashba, G E Onishchenko
ABSTRACT
Entosis is a type of cell cannibalism during which one cell penetrates into another cell and usually dies inside it. Researchers mainly pay attention to initial and final stages of entosis. Besides, tumor cells in suspension are the primary object of studies. In the present study, we investigated morphological changes of both cells-participants of entosis during this process. The substrate-dependent culture of human normal keratinocytes HaCaT was chosen for the work. A combination of light microscopy and scanning electron microscopy was used to prove that one cell was completely surrounded by the plasma membrane of another cell. We investigated such "cell-in-cell" structures and described the structural and functional changes of both cells during entosis. The outer cell nucleus localization and shape were changed. Gradual degradation of the inner cell nucleus and of the junctions between the inner and the outer cells was revealed. Moreover, repeated redistribution of the outer cell membrane organelles (Golgi apparatus, lysosomes, mitochondria, and autophagosomes), rearrangement of its cytoskeleton, and change in the lysosomal, autophagosomal, and mitochondrial state in both entotic cells were observed during entosis. On the basis of these data, we divided entosis into five stages that make it possible to systematize description of this type of cell death.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Picric acid, moistened with water, ≥98%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Hydriodic acid, distilled, 57 wt. %, 99.999% trace metals basis
Sigma-Aldrich
Rhodamine 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture