Merck
  • Inhibition of TRPM7 Attenuates Rat Aortic Smooth Muscle Cell Proliferation Induced by Angiotensin II: Role of Genistein.
Inhibition of TRPM7 Attenuates Rat Aortic Smooth Muscle Cell Proliferation Induced by Angiotensin II: Role of Genistein.
Journal of cardiovascular pharmacology (2015-07-15)
Meimei Yang, Tingting Zhao, Jinghan Lin, Ting Ju, Liming Zhang
ABSTRACT
Transient receptor potential melastatin 7 (TRPM7) is a Ca, Mg permeable nonselective cation channel of the TRP channel superfamily and plays an important role in cell growth and proliferation. Compounds that alter the activity and expression of the channel protein might be of therapeutic interest. In this study, we investigated the effects of genistein on TRPM7 channels and the proliferation of rat aortic smooth muscle cells (RAoSMCs). In primary cultured RAoSMCs, acute genistein (50 μM) exposure inhibited native TRPM7 currents, whereas chronic expose to genistein (50 μM) downregulated TRPM7 protein expression. The downregulation of TRPM7 protein expression induced by genistein was mimicked by c-Src inhibitor (PP2), but not by epidermal growth factor receptor tyrosine kinase inhibitor (lavendustin A), or daidzein. Additionally, genistein (50 μM) attenuated angiotensin II-induced cell proliferation. This study is the first to demonstrate inhibition of TRPM7 by isoflavone genistein through c-Src tyrosine kinase inhibition in RAoSMCs. Our results not only provide a new modulation mechanism of TRPM7 but also suggest that TRPM7 may serve as a new therapeutic target of genistein in the treatment of vascular diseases.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Pyrimidine, ≥98.0%