Merck
  • Mechanism of Dose-Dependent Regulation of UBE1L by Polyphenols in Human Bronchial Epithelial Cells.
Mechanism of Dose-Dependent Regulation of UBE1L by Polyphenols in Human Bronchial Epithelial Cells.
Journal of cellular biochemistry (2015-02-05)
Apei Jiang, Yuan Li, Pengqi Wang, Xiaoyun Shan, Pan Jiang, Xuemin Wang, Qing Feng
ABSTRACT
Ubiquitin activating enzyme E1-like (UBE1L) is the activating enzyme for ISG15ylation (ISG15, interferon stimulated gene 15). UBE1L is thought to be a candidate tumor suppressor gene and has positive activity against stress responses such as viral infections. Both type I interferon and retinoic acid are known to induce UBE1L expression. However, the molecular mechanism of UBE1L regulation is unclear. Here, the effect of several chemopreventive polyphenols on UBE1L expression in human bronchial epithelial cells (Beas-2B) was investigated. Lower concentrations of curcumin, (-)-epigallocatechin-3-gallate (EGCG) and resveratrol upregulated UBE1L, while high concentrations of curcumin, EGCG and resveratrol downregulated UBE1L levels. Interestingly, curcumin, EGCG and resveratrol diminished intracellular reactive oxygen species (ROS) at lower concentrations but generated ROS at higher concentrations. The antioxidant N-acetylcysteine (NAC) increased UBE1L protein levels, while pro-oxidants such as hydrogen peroxide and tert-butyl hydroperoxide (tBHP) decreased UBE1L protein levels, indicating that the intracellular redox status is associated with UBE1L expression. Kinase inhibitors were used to examine the contribution of mitogen-activated protein kinase (MAPK) activity to the polyphenol-regulated UBE1L. Only the inhibition of c-Jun N-terminal kinase (JNK) significantly reduced UBE1L expression. Knockdown of nuclear factor erythroid-2 related factor-2 (Nrf2) caused a concomitant decrease in UBE1L protein levels. It is concluded from the above mentioned results that JNK/Nrf2 signal pathway is involved in the regulation of UBE1L via intracellular ROS status when cells came in contact with polyphenols.
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
MTT Formazan, MTT reduction end product
Sigma-Aldrich
MTT Formazan, powder
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture