Merck
  • Association of Serotonin Transporter Promoter Polymorphism (5-HTTLPR) with Microscopic Colitis and Ulcerative Colitis.
Association of Serotonin Transporter Promoter Polymorphism (5-HTTLPR) with Microscopic Colitis and Ulcerative Colitis.
Digestive diseases and sciences (2014-12-24)
Arbab Sikander, Saroj Kant Sinha, Kaushal Kishor Prasad, Satya Vati Rana
ABSTRACT
Serotonin (5-HT) release and serotonin reuptake transporter (5-HTT) expression have been reported to be decreased in experimental colitis, in interleukin-10 knockout-associated colitis, and in patients with ulcerative colitis. Serotonin is known to play an important role in the pathogenesis of colitis, but individual genetic variants of 5-HTT gene in microscopic colitis and ulcerative colitis are not known. This study aimed to evaluate the association between the serotonin transporter gene promoter polymorphism (5-HTTLPR) and 5-HT concentration in microscopic colitis (MC) and ulcerative colitis (UC) patients. This prospective case-control study included 41 patients with microscopic colitis (age 19-82 years, mean 35 ± 13.6), 75 patients with ulcerative colitis (age 16-65 years, mean 38.5 ± 11.6), and 100 controls (age 20-64 years, mean 38 ± 11). 5-HTTLPR gene polymorphism was studied by polymerase chain reaction-based assay. 5-HT levels were measured by ELISA. The frequency of the 5-HTTLPR (SS) genotype was significantly lower in MC (12 %) patients compared to controls (30 %) (p < 0.05). When the L/L and L/S genotypes were combined into one group, the frequencies of the non-S genotype were significantly higher than those of S/S genotype between the MC patients and the controls (p < 0.05). 5-HT levels were significantly higher in UC and MC patients compared to healthy controls (p < 0.01). A significant association was observed between LL genotype of 5-HTTLPR polymorphism and microscopic colitis, suggesting that 5-HTTLPR is a potential candidate gene involved in the pathogenesis of microscopic colitis. Serotonin levels were significantly higher in microscopic colitis and ulcerative colitis patients compared to healthy controls.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent