- Role of textile substrate hydrophobicity on the adsorption of hydrosoluble nonionic block copolymers.
Role of textile substrate hydrophobicity on the adsorption of hydrosoluble nonionic block copolymers.
Journal of colloid and interface science (2015-05-26)
Junlong Song, Carlos Salas, Orlando J Rojas
PMID26004573
ABSTRACT
The adsorption of polyalkylene glycols and co-polymers of ethylene oxide and propylene oxide on substrates relevant to textiles with varying surface energies (cellulose, polypropylene, nylon and polyester) was studied by using quartz crystal microgravimetry. Langmuirian-type isotherms were observed for the adsorption profiles of nonionic block polymers of different architectures. The affinity with the surfaces is discussed based on experimental observations, which highlights the role of hydrophobic effects. For a given type of block polymer, micellar and monomeric adsorption is governed by the balance of polymer structure (mainly, chain length of hydrophobic segments) and substrate's surface energy.
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max™, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)