Merck
  • Using carboxyfluorescein diacetate succinimidyl ester to monitor intracellular protein glycation.
Using carboxyfluorescein diacetate succinimidyl ester to monitor intracellular protein glycation.
Analytical biochemistry (2015-03-25)
Julie Boucher, Élie Simard, Ulrike Froehlich, Pedro D'Orléans-Juste, Michel Grandbois
ABSTRACT
Protein glycation is a ubiquitous process involved in vascular complications observed in diabetes. Glyoxal (GO), an intracellular reactive oxoaldehyde that is one of the most potent glycation agents, readily reacts with amines present on proteins to produce the lysine-derived adduct carboxymethyllysine, which is a prevalent advanced glycation end-product (AGE). Our group previously showed that cell exposure to GO leads to an alteration in the cell contractile activity that could occur as a result of the glycation of various proteins regulating the cell contractile machinery. Here, we measured the extent of glycation on three functionally distinct proteins known to participate in cell contraction and cytoskeletal organization-Rho-kinase (ROCK), actin, and gelsolin (GSN)-using an assay based on the reaction of the cell membrane-permeable fluorescent probe carboxyfluorescein diacetate succinimidyl ester (CFDA-SE), which reacts with primary amine groups of proteins. By combining CFDA-SE fluorescence and Western blot detection, we observed (following GO incubation) increased glycation of actin and ROCK as well as an increased interaction between actin and GSN as observed by co-immunoprecipitation. Thus, we conclude that the use of the fluorescent probe CFDA-SE offers an interesting alternative to perform a comparative analysis of the extent of intracellular protein glycation in live cells.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
4-Nitrophenyl acetate, esterase substrate
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
SAFC
L-Glutamine