Merck
  • miR-29b attenuates tumorigenicity and stemness maintenance in human glioblastoma multiforme by directly targeting BCL2L2.
miR-29b attenuates tumorigenicity and stemness maintenance in human glioblastoma multiforme by directly targeting BCL2L2.
Oncotarget (2015-07-15)
Hyun Joo Chung, Young Eun Choi, Eun Sook Kim, Young-Hoon Han, Myung-Jin Park, In Hwa Bae
ABSTRACT
Glioblastoma multiforme (GBM) is the most common malignant brain tumor and exhibits aggressive and invasive behavior. We previously identified four miRNAs-miR-29b, 494, 193a-3p, and 30e-with enhanced expression in GBM following treatment of ionizing radiation by miRNA microarray analysis. In this study, we found that only miR-29b inhibited tumor cell migration and invasion by reducing MMP-2 activity via phospho-AKT/β-catenin signaling, and stimulated a more epithelial-like morphology. Moreover, miR-29b inhibits angiogenesis by attenuating tube formation and the expression of VEGF and Ang-2, and stemness maintenance in GBM cells, as demonstrated by decreasing neurosphere formation and cancer stem cell marker protein expression. These findings support the anti-tumor properties of miR-29b in human GBM cells. Furthermore, miR-29b expression was inversely proportional to that of BCL2L2 mRNA or protein in various cancer cell types. Interestingly, BCL2L2 mRNA is highly expressed in the mesenchymal type of GBM. To further elucidate the relationship between miR-29b and BCL2L2 in GBM, we performed co-transfection reporter assays and determined that miR-29b downregulates BCL2L2 expression by directly binding its 3'UTR. Finally, we confirmed that BCL2L2 repression is of central importance to miR-29b anti-tumor activity using functional assays to examine cell migration, invasion, angiogenesis, and stemness. From these data, we propose that miR-29b may be a useful therapeutic agent in GBM.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bcl2l2
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
MISSION® esiRNA, targeting human BCL2L2