Merck
  • Multi-region to single region shear thinning transitions in drying PEDOT:PSS dispersions: contributions from charge density fluctuations.
Multi-region to single region shear thinning transitions in drying PEDOT:PSS dispersions: contributions from charge density fluctuations.
Soft matter (2015-09-12)
Piramuthu Raja Ashok R, Mathew Shaji Thomas, Susy Varughese
ABSTRACT
The multiple shear thinning behaviour observed in dispersions of the conducting polymer, poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate) ( PSS), in water during the drying process is explored. PSS dispersions in water can be non-stoichiometric polyelectrolyte complexes (PECs) with free polyanion (PSS) chains resulting in scrambled egg conformations. They also behave as charged colloidal particles. The behaviour of PSS dispersions as PECs and charged colloidal systems during drying is analyzed using rheology and TEM, zeta potential, conductivity, pH, dynamic light scattering (DLS) and rheo-small angle light scattering (rheo-SALS) information. The rheological behaviour is found to have significant contributions from conformational changes in the PSS PEC due to charge density fluctuations affecting the electrostatic persistence length, le. A low concentration regime with multiple shear thinning behaviour and a high concentration regime with single shear thinning behaviour were observed. The PEC behavior of PSS is further explored by charge neutralizing the dispersions with salts.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis