Merck
  • Gonadotrophin-releasing hormone agonists for fertility preservation: unraveling the enigma?
Gonadotrophin-releasing hormone agonists for fertility preservation: unraveling the enigma?
Human reproduction (Oxford, England) (2015-03-23)
Noa Hasky, Shiri Uri-Belapolsky, Keren Goldberg, Irit Miller, Hadas Grossman, Salomon M Stemmer, Irit Ben-Aharon, Ruth Shalgi
ABSTRACT
Can gonadotrophin-releasing hormone agonists (GnRH-a) preserve long-term fertility when administered prior to and concomitantly with chemotherapy? GnRH-a display a differential protective effect on fertility, depending upon the specific chemotherapy-induced mechanism of ovarian injury. The role of GnRH-a in fertility preservation has been constantly debated and their use is considered experimental due to conflicting clinical evidence and paucity of data regarding their mechanism for ovarian protection. In vivo model: 7-8 weeks old imprinting control region (ICR) mice were injected with GnRH-a (Leuprolide-acetate) or saline prior to and concomitantly with cyclophosphamide, doxorubicin or saline and sacrificed at various time-points on a longitudinal follow-up; 24 h (n = 36), 1 week (n = 40), 1 month (n = 36) and 9 months (n = 66) post chemotherapy treatment. Blood samples were drawn on Day 0 and on a monthly basis after chemotherapy treatment. On the day of sacrifice, blood samples were drawn and ovaries excised and processed for either immunohistochemistry (IHC), protein or RNA extraction. In vitro model: 21-23 days old Wistar-derived rats were sacrificed, their ovaries excised and primary granulosa cells (PGC) were either isolated for in vitro culture, or processed for immunofluorescence (IF) as well as for protein or RNA extraction. Ovarian reserve was estimated by serial measurements of serum anti-mullerian hormone (AMH), quantified by the AMH Gen II ELISA assay. Ovarian AMH and phosphorylated Akt (pAkt) were detected by immunoblotting. Vascular endothelial growth factor (VEGF) was measured by quantitative PCR. Ovarian GnRH receptor (GnRHR), AMH and CD34 were visualized by IHC, and apoptosis was evaluated using TdT (terminal deoxynucleotidyl transferase)-mediated dUDP nick-end labeling (TUNEL). Cyclophosphamide-induced ovarian injury caused a prompt decrease in AMH level (P < 0.01) and a further long-term decline in serum AMH (P = 0.017), indicating damage to the ovarian reserve. Pretreatment with GnRH-a diminished AMH-decrease (P < 0.05) and maintained serum AMH level in the long run (P < 0.05). Doxorubicin-exerted ovarian-vascular-injury is also displayed by an acute increase in ovarian VEGF level (P < 0.05) and a sustained decrease in serum AMH level (P < 0.001). This was followed by ovarian recovery manifested by increased neovascularization. GnRH-a delayed the recovery in AMH level and decreased the level of VEGF (P < 0.001), thus interfering with the vascular recovery subsequent to doxorubicin-induced vascular damage. To portray the differential mechanism of each chemotherapy, cyclophosphamide and doxorubicin were given separately, whereas most of the clinical protocols include several types of chemotherapies. Thus, future study should explore a prospective evaluation of various chemotherapies, as well as combined chemotherapeutic protocols. Our study demonstrates that different chemotherapy agents affect the ovary via diverse mechanisms and thus the administration of GnRH-a concomitantly, could be beneficial to a subpopulation of patients treated with cyclophosphamide-based protocols. This work was partially supported by a grant from the Israel Science Foundation (ISF) to I.B.-A. The authors have no conflict of interest to disclose.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)