Merck
  • Mutation of a Single Envelope N-Linked Glycosylation Site Enhances the Pathogenicity of Bovine Leukemia Virus.
Mutation of a Single Envelope N-Linked Glycosylation Site Enhances the Pathogenicity of Bovine Leukemia Virus.
Journal of virology (2015-06-19)
Alix de Brogniez, Amel Baya Bouzar, Jean-Rock Jacques, Jean-Philippe Cosse, Nicolas Gillet, Isabelle Callebaut, Michal Reichert, Luc Willems
ABSTRACT
Viruses have coevolved with their host to ensure efficient replication and transmission without inducing excessive pathogenicity that would indirectly impair their persistence. This is exemplified by the bovine leukemia virus (BLV) system in which lymphoproliferative disorders develop in ruminants after latency periods of several years. In principle, the equilibrium reached between the virus and its host could be disrupted by emergence of more pathogenic strains. Intriguingly but fortunately, such a hyperpathogenic BLV strain was never observed in the field or designed in vitro. In this study, we sought to understand the role of envelope N-linked glycosylation with the hypothesis that this posttranslational modification could either favor BLV infection by allowing viral entry or allow immune escape by using glycans as a shield. Using reverse genetics of an infectious molecular provirus, we identified a N-linked envelope glycosylation site (N230) that limits viral replication and pathogenicity. Indeed, mutation N230E unexpectedly leads to enhanced fusogenicity and protein stability. Infection by retroviruses requires the interaction of the viral envelope protein (SU) with a membrane-associated receptor allowing fusion and release of the viral genomic RNA into the cell. We show that N-linked glycosylation of the bovine leukemia virus (BLV) SU protein is, as expected, essential for cell infection in vitro. Consistently, mutation of all glycosylation sites of a BLV provirus destroys infectivity in vivo. However, single mutations do not significantly modify replication in vivo. Instead, a particular mutation at SU codon 230 increases replication and accelerates pathogenesis. This unexpected observation has important consequences in terms of disease control and managing.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
SAFC
L-Glutamine
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)