Merck
  • Pharmacology of smac mimetics; chemotype differentiation based on physical association with caspase regulators and cellular transport.
Pharmacology of smac mimetics; chemotype differentiation based on physical association with caspase regulators and cellular transport.
Experimental cell research (2015-08-25)
Randy L Talbott, Robert M Borzilleri, Charu Chaudhry, Joseph Fargnoli, Henry Shen, Craig Fairchild, Bryan Barnhart, Marie Ortega, Thomas E McDonagh, Ragini Vuppugalla, Gregory D Vite, John T Hunt, Marco Gottardis, Joseph G Naglich
ABSTRACT
Cellular levels of inhibitor of apoptosis (IAP) proteins are elevated in multiple human cancers and their activities often play a part in promoting cancer cell survival by blocking apoptotic pathways, controlling signal transduction pathways and contributing to resistance. These proteins function through interactions of their BIR (baculoviral IAP repeat) protein domains with pathway components and these interactions are endogenously antagonized by Smac/Diablo (second mitochondrial activator of caspases/direct IAP binding protein with low isoelectric point). This report describes development of synthetic smac mimetics (SM) and compares their binding, antiproliferative and anti-tumor activities. All dimeric antagonists inhibit in vitro smac tetrapeptide binding to recombinant IAP proteins, rescue IAP-bound caspase-3 activity and show anti-proliferative activity against human A875 melanoma cells. One heterodimeric SM, SM3, binds tightly to IAP proteins in vitro and slowly dissociates (greater than two hours) from these protein complexes compared to the other antagonists. In addition, in vitro SM anti-proliferation potency is influenced by ABCB1 transporter (ATP-binding cassette, sub-family B; MDR1, P-gp) activities and one antagonist, SM5, does not appear to be an ABCB1 efflux pump substrate. All dimeric smac mimetics inhibit the growth of human melanoma A875 tumors implanted in athymic mice at well-tolerated doses. One antagonist, SM4, shows broad spectrum in vivo anti-tumor activity and modulates known pharmacodynamic markers of IAP antagonism. These data taken together demonstrate the range of diverse dimeric IAP antagonist activities and supports their potential as anticancer agents.
Sigma-Aldrich
6-Carboxyfluorescein, ≥96% (HPLC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
6-Maleimidohexanoic acid, 90% (GC)
Terbium, powder, 5g, max. particle size 500 micron, 99.9%
Terbium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Terbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Terbium, powder, 2g, max. particle size 500 micron, 99.9%
Terbium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Terbium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, ≤1.0 mol % L-α-isomer
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Glycerol phosphate disodium salt hydrate, isomeric mixture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)