Merck
  • A first survey on the biochemical composition of egg yolk and lysozyme-like activity of egg envelopment in the cuttlefish Sepia officinalis from the Northern Adriatic Sea (Italy).
A first survey on the biochemical composition of egg yolk and lysozyme-like activity of egg envelopment in the cuttlefish Sepia officinalis from the Northern Adriatic Sea (Italy).
Fish & shellfish immunology (2015-05-20)
Valerio Matozzo, Irene Conenna, Verena Maria Riedl, Maria Gabriella Marin, Tihana Marčeta, Carlotta Mazzoldi
ABSTRACT
The cuttlefish Sepia officinalis is an important fishery resource in the Northern Adriatic Sea (Italy). During reproduction, fertilised eggs are released by adult females in coastal waters and embryo development can take over two months. During this period, embryos rely on nutrients and other substances, such as immune factors, provided by the female in egg yolk. In cephalopods in general, and specifically in the common cuttlefish, little information is available on yolk biochemical composition and substances included in egg envelopment. In the present study, the main biochemical components of egg yolk and the presence of antimicrobial substances in egg envelopment of S. officinalis were determined for the first time. Statistically significant differences in total egg weight and egg yolk weight were observed among batches from different females. Egg and yolk weights were positively correlated, with yolk representing the 13% (±5%) of the total egg weight. Total proteins were the main biochemical component (46%) of egg yolk, followed by total carbohydrates plus glycogen (39%) and lipids (15%). Statistically significant differences among batches were recorded in egg yolk total protein amounts, lipids, carbohydrates and glycogen, but no correlations were found between egg yolk weight and the biochemical components. The Petri dish and the quantitative spectrophotometric assays revealed the presence of lysozyme-like activity in egg gelatinous envelopment.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene blue, certified by the BSC