Merck
  • Epitope characterization of the ADA response directed against a targeted immunocytokine.
Epitope characterization of the ADA response directed against a targeted immunocytokine.
Journal of pharmaceutical and biomedical analysis (2015-06-22)
Kay Stubenrauch, Christian Künzel, Rudolf Vogel, Dietrich Tuerck, Eginhard Schick, Julia Heinrich
ABSTRACT
Targeted immunocytokines (TICs) display potent activity in selective tumor suppression. This class of multi domain biotherapeutics (MDBs) is composed of the three major domains Fab, Fc, and a cytokine which may induce a complex polyclonal anti-drug antibody (ADA) response. However, classical ADA assays usually are not suitable to specify ADAs and to identify the immunogenic domains of a TIC. The purpose of the present study was to establish epitope characterization of ADA responses in order to specify immunogenic responses against a TIC and their direct impact on the pharmacokinetic profile, safety, and efficacy. Based on standard ADA screening and confirmation assays, respectively, domain detection assays (DDAs) and domain competition assays (DCAs) were established and compared by the use of 12 ADA-positive samples obtained from a cynomolgus monkey study in early development. Both domain-specific assays were sensitive enough to preserve the positive screening assay result and revealed an overall accordance for the evaluation of domain-specific ADA responses. About half of the samples displayed one ADA specificity, either for the Fab or for the cytokine (Cy) domain, and the remaining samples showed a combination of Fab-specific and Cy-specific ADA fractions. Fc-specific ADAs occurred in only one sample. In-depth comparison of DCAs and DDAs showed that both assays appeared to be appropriate to assess multi-specific ADA responses as well as minor ADA fractions. An advantage of DCAs is typically a fast and easy assay establishment, whereas, DDAs in some cases may be superior to assess low abundant ADAs in multi-specific responses. Our results reveal that both approaches benefit from thorough reagent development as an essential precondition for reliable epitope characterization of ADA responses.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
Iodoacetamide
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
N,N-Dimethyldodecylamine, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis