Merck
  • Bergamot polyphenol fraction prevents nonalcoholic fatty liver disease via stimulation of lipophagy in cafeteria diet-induced rat model of metabolic syndrome.
Bergamot polyphenol fraction prevents nonalcoholic fatty liver disease via stimulation of lipophagy in cafeteria diet-induced rat model of metabolic syndrome.
The Journal of nutritional biochemistry (2015-05-31)
Maddalena Parafati, Antonella Lascala, Valeria Maria Morittu, Francesca Trimboli, Antonia Rizzuto, Elvira Brunelli, Francesca Coscarelli, Nicola Costa, Domenico Britti, James Ehrlich, Ciro Isidoro, Vincenzo Mollace, Elzbieta Janda
ABSTRACT
Nonalcoholic fatty liver disease (NAFLD) is the most common liver disease in industrialized countries. Defective autophagy of lipid droplets (LDs) in hepatocytes, also known as lipophagy, has recently been identified as a possible pathophysiological mechanism of NAFLD. Experimental and epidemiological evidence suggests that dietary polyphenols may prevent NAFLD. To address this hypothesis and analyze the underlying mechanisms, we supplemented bergamot polyphenol fraction (BPF) to cafeteria (CAF) diet-fed rats, a good model for pediatric metabolic syndrome and NAFLD. BPF treatment (50 mg/kg/day supplemented with drinking water, 3 months) potently counteracted the pathogenic increase of serum triglycerides and had moderate effects on blood glucose and obesity in this animal model. Importantly, BPF strongly reduced hepatic steatosis as documented by a significant decrease in total lipid content (-41.3% ± 12% S.E.M.), ultrasound examination and histological analysis of liver sections. The morphometric analysis of oil-red stained sections confirmed a dramatic reduction in LDs parameters such as total LD area (48.5% ± 15% S.E.M.) in hepatocytes from CAF+BPF rats. BPF-treated livers showed increased levels of LC3 and Beclin 1 and reduction of SQSTM1/p62, suggesting autophagy stimulation. Consistent with BPF stimulation of lipophagy, higher levels of LC3II were found in the LD subcellular fractions of BPF-expose livers. This study demonstrates that the liver and its lipid metabolism are the main targets of bergamot flavonoids, supporting the concept that supplementation of BPF is an effective strategy to prevent NAFLD.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
SAFC
BIS-TRIS