Merck
  • Development and validation of a rapid test for anaerobic inhibition and toxicity.
Development and validation of a rapid test for anaerobic inhibition and toxicity.
Water research (2015-06-13)
S Astals, D J Batstone, S Tait, P D Jensen
ABSTRACT
Despite the importance of quantifying inhibitory capacity of compounds in anaerobic digestion, there is currently no well-defined method to assess it. Experimental methods in literature are frequently time-consuming and resource intensive. As a result, detailed inhibition testing rarely forms part of anaerobic digestion studies, despite the importance and utility of this information. This study develops and validates a simple and rapid inhibition test protocol, based on relative inhibition of acetoclastic methanogens. The inhibition potential of a compound is determined from the reduction in specific methanogenic activity as inhibitor concentration is increased. The method was successfully performed on two inoculums from different source environments and with both biostatic and biocidal inhibitors. Optimisation work indicated that: (i) sodium acetate is a preferred carbon source compared to acetic acid; (ii) an inoculum to acetate ratio of 5 g VS g(-1) acetate is preferred, and (iii) that the inoculum concentration should be normalised to 10 g L(-1) VS to reduce mass transfer problems and promote consistency. A key advantage over existing methods is that the sampling strategy has been optimised to three events over 1.5 days while effectively controlling the relative analytical error.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)