Merck
  • Preparation of malathion MIP-SPE and its application in environmental analysis.
Preparation of malathion MIP-SPE and its application in environmental analysis.
Environmental monitoring and assessment (2015-06-04)
Hai Gen Zuo, Jian Xin Zhu, Chun Rui Zhan, Lei Shi, Ming Xing, Ping Guo, Yuan Ding, Hong Yang
ABSTRACT
Malathion is an organophosphorous insecticide for controlling insects on fruits and vegetables, miscellaneous household insects, and animal parasites. It is important to develop highly efficient and selective pre-treatment method for analyzing malathion residues in environment and samples from agricultural products based on the molecularly imprinted polymers (MIPs). In this study, we developed a tailor-made MIP method with highly specific recognization to the template. The MIPs were prepared using malathion as a template, methacrylic acid (MAA) as a functional monomer, ethylene glycol dimethacrylate (EGDMA) as a crosslinker, azodiisobutyronitrile (AIBN) as an initiator, and the acetonitrile-chloroform (1:1, v/v) as a porogen. The molecular recognization mechanism of malathion and MAA was evaluated by molecular simulation, ultraviolet spectrometry (UV), and (1)H-nuclear magnetic resonance ((1)H-NMR). MAA interacted specifically with malathion by hydrogen bond with a ratio of 2:1. The MIPs exhibit a high affinity, recognition specificity, and efficient adsorption performance for malathion. The Fourier transform infrared spectroscopy (FT-IR), scanning electron microscope (SEM), surface area and porosimeter analyzer, thermogravimetric/differential thermal analyzer (TG/DTA) were used to characterize the properties of MIP. The malathion residues in soil, tap water, and cabbage were cleaned up by MIP-SPE, detected quantitatively using GC-FPD, and confirmed by GC-MS/MS. The limits of tap water, soil, and cabbage were confined to 0.001 mg L(-1), 0.004 and 0.004 mg kg(-1), respectively. The spiked recoveries of malathion were 96.06-111.49% (with RSD being 5.7-9.2%), 98.13-103.83% (RSD, 3.5-8.7%), and 84.94-93.69% (RSD, 4.7-5.8%) for tap water, soil, and cabbage samples, respectively. Thus, the method developed here can be used effectively in assessing malathion residues in multiple environmental samples. The aim of the study was to provide an efficient, selective, and accurate method for analyzing malathion at trace levels in multiple media.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis