Merck
  • Orally administrated pterostilbene attenuates acute cerebral ischemia-reperfusion injury in a dose- and time-dependent manner in mice.
Orally administrated pterostilbene attenuates acute cerebral ischemia-reperfusion injury in a dose- and time-dependent manner in mice.
Pharmacology, biochemistry, and behavior (2015-06-19)
Yu Zhou, Xue-mei Zhang, Ang Ma, Ya-li Zhang, Yan-yi Chen, Hao Zhou, Wen-jun Li, Xin Jin
ABSTRACT
Pterostilbene (3,5-dimethoxy-4-hydroxystilbene) is a component of blueberry. It has been reported that long-term treatment with blueberry has a neuroprotective effect. However, it has not been reported whether pterostilbene is effective in attenuating cerebral ischemia/reperfusion (I/R) injury. In the present study, focal cerebral ischemia was induced by middle cerebral artery occlusion for 90min followed by reperfusion. To observe the dose-dependent effect, pterostilbene (2.5-80mg/kg, ig) was administered for 3days before ischemia. To determine the time-dependent effect, pterostilbene (10mg/kg, ig) was administered as a single dose at 0, 1, or 3h after reperfusion. Twenty-four hours after I/R, pterostilbene dose-dependently improved neurological function, reduced brain infarct volume, and alleviated brain edema. The most effective dose was 10mg/kg; the therapeutic time window was within 1h after I/R and treatment immediately after reperfusion showed the best protective effect. The protective effect is further confirmed by the results that post-ischemic treatment with pterostilbene (10mg/kg) significantly improved motor function, alleviated blood brain barrier disruption, increased neurons survival and reduced cell apoptosis in cortical penumbra after cerebral I/R. We also found that pterostilbene (10mg/kg) significantly reversed the increased content of malondialdehyde and the decreased activity of superoxide dismutase in the ipsilateral hemisphere. Furthermore, pterostilbene decreased the oxidative stress markers 4-hydroxynonenal and 8-hydroxyguanosine positive cells in the cortical penumbra. All these findings indicate that pterostilbene dose- and time-dependently exerts a neuroprotective effect against acute cerebral I/R injury. This neuroprotective effect of pterostilbene may be associated with its inhibition of oxidative stress and subsequent neuronal apoptosis in the cortical penumbra.
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, ≥98.0% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Pterostilbene, ≥97% (HPLC), solid
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N