Merck
  • Neisseria meningitidis Group A IgG1 and IgG2 Subclass Immune Response in African Children Aged 12-23 Months Following Meningococcal Vaccination.
Neisseria meningitidis Group A IgG1 and IgG2 Subclass Immune Response in African Children Aged 12-23 Months Following Meningococcal Vaccination.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2015-11-11)
Daniel Holme, Helen Findlow, Samba O Sow, Olubukola T Idoko, Marie-Pierre Preziosi, George Carlone, Brian D Plikaytis, Ray Borrow
ABSTRACT
A group A meningococcal conjugate vaccine, PsA-TT, was licensed in 2010 and was previously studied in a phase 2 clinical trial to evaluate its safety and immunogenicity in African children 12-23 months of age. Subjects received either PsA-TT; meningococcal group A, C, W, Y polysaccharide vaccine (PsACWY); or Haemophilus influenzae type b conjugate vaccine (Hib-TT). Forty weeks following primary vaccination, the 3 groups were further randomized to receive either PsA-TT, one-fifth dose of PsACWY, or Hib-TT. Group A-specific immunoglobulin G (IgG) subclass response was characterized using an enzyme-linked immunosorbent assay. The predominant IgG subclass response, regardless of vaccine, was IgG1. One month following primary vaccination, the geometric mean concentrations (GMCs) of IgG1 and IgG2 in the PsA-TT group were 21.73 µg/mL and 6.27 µg/mL, whereas in the PsACWY group the mean GMCs were 2.01 µg/mL and 0.97 µg/mL, respectively (P < .0001). Group A-specific IgG1 and IgG2 GMCs remained greater in the PsA-TT group than in the PsACWY group 40 weeks following primary vaccination (P < .0001). One week following revaccination, those given 2 doses of PsA-TT had the greatest IgG1 and IgG2 GMCs of 125.23 µg/mL and 36.12 µg/mL, respectively (P = .0008), and demonstrated a significant increase in IgG1:IgG2 mean ratio, indicative of the T-cell-dependent response associated with conjugate vaccines. Vaccination of African children aged 12-24 months with either PsA-TT or PsACWY elicited a predominantly IgG1 response. The IgG1:IgG2 mean ratio decreased following successive vaccination with PsACWY, indicating a shift toward IgG2, suggestive of the T-cell-independent immune response commonly associated with polysaccharide antigens. SRCTN78147026.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Aluminum phosphate, 99.99% trace metals basis
Sigma-Aldrich
Ethylamine, 97%
Sigma-Aldrich
Aluminum phosphate, reagent grade
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylamine solution, 66.0-72.0% in H2O
Sigma-Aldrich
Ethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Ethylamine solution, 2.0 M in THF