Merck
  • Recapitulation and Modulation of the Cellular Architecture of a User-Chosen Cell of Interest Using Cell-Derived, Biomimetic Patterning.
Recapitulation and Modulation of the Cellular Architecture of a User-Chosen Cell of Interest Using Cell-Derived, Biomimetic Patterning.
ACS nano (2015-05-20)
John H Slater, James C Culver, Byron L Long, Chenyue W Hu, Jingzhe Hu, Taylor F Birk, Amina A Qutub, Mary E Dickinson, Jennifer L West
ABSTRACT
Heterogeneity of cell populations can confound population-averaged measurements and obscure important findings or foster inaccurate conclusions. The ability to generate a homogeneous cell population, at least with respect to a chosen trait, could significantly aid basic biological research and development of high-throughput assays. Accordingly, we developed a high-resolution, image-based patterning strategy to produce arrays of single-cell patterns derived from the morphology or adhesion site arrangement of user-chosen cells of interest (COIs). Cells cultured on both cell-derived patterns displayed a cellular architecture defined by their morphology, adhesive state, cytoskeletal organization, and nuclear properties that quantitatively recapitulated the COIs that defined the patterns. Furthermore, slight modifications to pattern design allowed for suppression of specific actin stress fibers and direct modulation of adhesion site dynamics. This approach to patterning provides a strategy to produce a more homogeneous cell population, decouple the influences of cytoskeletal structure, adhesion dynamics, and intracellular tension on mechanotransduction-mediated processes, and a platform for high-throughput cellular assays.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline