Merck
  • Microminipigs as a new experimental animal model for toxicological studies: comparative pharmacokinetics of perfluoroalkyl acids.
Microminipigs as a new experimental animal model for toxicological studies: comparative pharmacokinetics of perfluoroalkyl acids.
Journal of applied toxicology : JAT (2015-04-17)
Keerthi S Guruge, Michiko Noguchi, Koji Yoshioka, Eriko Yamazaki, Sachi Taniyasu, Miyako Yoshioka, Noriko Yamanaka, Mitsutaka Ikezawa, Nobuhiko Tanimura, Masumi Sato, Nobuyoshi Yamashita, Hiroaki Kawaguchi
ABSTRACT
In this study, we evaluated the efficacy of a novel minipig strain, the Microminipig (MMPig), as an animal model for studying the pharmacokinetics of a mixture of 10 perfluoroalkyl acids (PFAAs). After a single oral dose was given, we found that the blood depuration of PFAAs (blood t1/2), which we calculated using first-order elimination curves, ranged from 1.6 to 86.6 days. Among the five body compartments analyzed, the liver was the greatest site of accumulation of perfluorooctanesulfonate and longer chain perfluorinated carboxylates such as perfluorodecanoic acid, perfluoroundecanoic acid and perfluorododecanoic acid. We observed an increasing accumulation trend of perfluorinated carboxylates in the organs associated with the fluorinated carbon chain length. The perfluorononanoic acid burden was the highest among the treated compounds 21 days after a single exposure, as 29% of the given perfluorononanoic acid dose was accumulated in the tissues. The persistence of PFAAs in edible pig tissues even after 21 days post-exposure raises concerns about the safety of swine products. This was the first study to use MMPigs to elucidate the pharmacokinetics of a group of environmental pollutants. We found that MMPigs could be excellent experimental animals for toxicological studies due to their easy handling, cost efficacy for target compounds and ease of waste treatment.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Undecafluorohexanoic acid, ≥97.0% (T)
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.