Merck
  • Multiparametric assay using HepaRG cells for predicting drug-induced liver injury.
Multiparametric assay using HepaRG cells for predicting drug-induced liver injury.
Toxicology letters (2015-05-03)
Takafumi Tomida, Hayao Okamura, Masahiro Satsukawa, Tsuyoshi Yokoi, Yoshihiro Konno
ABSTRACT
The utility of HepaRG cells as an in vitro cell-based assay system for assessing drug-induced liver injury (DILI) risk was investigated. Seventeen DILI-positive and 15 DILI-negative drugs were selected for the assay. HepaRG cells were treated with each drug for 24h at concentrations that were 1.6-, 6.3-, 25-, and 100-fold the therapeutic maximum plasma concentration (Cmax). After treatment, the cell viability, glutathione content, caspase 3/7 activity, lipid accumulation, leakage of lactate dehydrogenase, and albumin secretion were measured. The sensitivity and specificity were calculated to assess the ability of the assay to predict DILI. Our multiparametric assay using HepaRG cells exhibited a 67% sensitivity and 73% specificity at a 100-fold concentration of Cmax and a 41% sensitivity and 87% specificity at a 25-fold concentration of Cmax. When a 25-fold Cmax cut-off was applied, approximately 70% of drugs exhibiting positive responses were classified into the high DILI risk category. HepaRG cells distinguished relatively safe drugs from their high-risk analogs. Our study indicates that HepaRG cells may be of use to (1) prioritize drug analogs, (2) analyze the mechanism of DILI, and (3) assess the risk for DILI in the early drug discovery stage.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Troglitazone, ≥98% (HPLC)
Sigma-Aldrich
Valsartan, ≥98% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
(±)-Miconazole nitrate salt, imidazole antibiotic
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Amiodarone hydrochloride, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Olanzapine, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Furosemide
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay), meets EP testing specifications
Sigma-Aldrich
3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt, ≥98%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%