Merck
  • Engineered Nanostructured Facial Lipopeptide as Highly Efficient Molecular Transporter.
Engineered Nanostructured Facial Lipopeptide as Highly Efficient Molecular Transporter.
ACS applied materials & interfaces (2015-08-05)
Chiranjit Dutta, Kasturee Chakraborty, Rituparna Sinha Roy
ABSTRACT
Designing an effective peptide based molecular transporter for the intracellular delivery of hydrophilic therapeutic biomacromolecules remains a considerable challenge. Highly basic oligoarginine and lipidated arginine rich cell penetrating peptides have been reported in the literature as molecular transporters, which were extensively used for cellular internalization of significantly large biopharmaceuticals. However, oligoarginine based molecular transporters with l-arginine residues pose significant challenges due to proteolytic instability and limited stability of noncovalent peptide-cargo nanocomplexes. Exploiting the rational peptide designing strategy, we have engineered protease-resistant facial lipopeptide based molecular transporter having arginine-sarcosine-arginine moiety to minimize adjacent arginine-arginine pair repulsion. N-Methylated amino acid sarcosine was incorporated as a spacer between two adjacent arginine residues, which provides proteolytic stability to the designed peptide and minimizes intermolecular aggregation of peptides. Two stearyl moieties were incorporated to facilitate cellular internalization. Interestingly, our designed lipopeptide exhibits significantly enhanced cellular internalization with only six l-arginine residues compared to stearylated oligo-nona-arginine. Additionally, enhanced proteolytic stability of such class of molecular transporter enables increased cargo internalization, and we anticipate that our engineered multifunctional, proteolytically stable, nanostructured facial lipopeptide based molecular transporter can have major impact in advancing drug delivery technologies.
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
(Benzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Benzotriazole, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer