Merck
  • Differential gene transcription, biochemical responses, and cytotoxicity assessment in Pacific oyster Crassostrea gigas exposed to ibuprofen.
Differential gene transcription, biochemical responses, and cytotoxicity assessment in Pacific oyster Crassostrea gigas exposed to ibuprofen.
Environmental science and pollution research international (2015-01-18)
Miguel A S Serrano, Maria Gonzalez-Rey, Jacó J Mattos, Fabrício Flores-Nunes, Álvaro C P Mello, Flávia L Zacchi, Clei E Piazza, Marília N Siebert, Rômi S Piazza, Diana Alvarez-Muñoz, Sara Rodriguez-Mozaz, Damià Barceló, Maria João Bebianno, Carlos H A M Gomes, Cláudio M R Melo, Afonso C D Bainy
ABSTRACT
Pharmaceuticals, such as anti-inflammatory nonsteroidal drugs, are frequently detected in aquatic ecosystems. Studies about the effects of these substances in nontarget organisms, such as bivalves, are relevant. The aim of this study was to evaluate the effects on antioxidant status caused by ibuprofen (IBU) in oysters Crassostrea gigas exposed for 1, 4, and 7 days at concentrations 1 and 100 μg L(-1). Levels of IBU in tissues of oysters, as well as cell viability of hemocytes, were measured. The transcription of cytochrome P450 genes (CYP2AU2, CYP356A1, CYP3071A1, CYP30C1), glutathione S-transferase isoforms (GST-ω-like and GST-π-like), cyclooxygenase-like (COX-like), fatty acid binding protein-like (FABP-like), caspase-like, heat shock protein-like (HSP70-like), catalase-like (CAT-like), and the activity of catalase (CAT), glutathione peroxidase (GPx), glutathione reductase (GR), and glutathione S-transferase (GST) were also evaluated in the gills of oysters. The highest levels of IBU were observed in animals exposed to 100 μg L(-1). A significant upregulation of CYP2AU1, CYP356A1, CYP3071A1, GST-ω-like, GST-π-like, COX-like, and FABP-like was observed in oysters exposed to IBU under different experimental conditions. Oysters exposed to 1 μg L(-1) for 7 days showed a significantly higher transcription of CYP2AU2, CYP356A1, CYP3071A1, GST-ω-like, and GST-π-like but lower GR activity. In conclusion, C. gigas exposed to environmentally relevant concentrations of IBU (1 μg L(-1)) exhibited increased transcription of certain genes and alterations on antioxidant and auxiliary enzymes, which could, in the the long term, cause damages to exposed organisms.
Product Description
Supelco
Ibuprofen
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~98%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ≥99.5%