Merck
  • Homozygous mdm2 SNP309 cancer cells with compromised transcriptional elongation at p53 target genes are sensitive to induction of p53-independent cell death.
Homozygous mdm2 SNP309 cancer cells with compromised transcriptional elongation at p53 target genes are sensitive to induction of p53-independent cell death.
Oncotarget (2015-09-30)
Melissa Rosso, Alla Polotskaia, Jill Bargonetti
ABSTRACT
A single nucleotide polymorphism (T to G) in the mdm2 P2 promoter, mdm2 SNP309, leads to MDM2 overexpression promoting chemotherapy resistant cancers. Two mdm2 G/G SNP309 cancer cell lines, MANCA and A875, have compromised wild-type p53 that co-localizes with MDM2 on chromatin. We hypothesized that MDM2 in these cells inhibited transcription initiation at the p53 target genes p21 and puma. Surprisingly, following etoposide treatment transcription initiation occurred at the compromised target genes in MANCA and A875 cells similar to the T/T ML-1 cell line. In all cell lines tested there was equally robust recruitment of total and initiated RNA polymerase II (Pol II). We found that knockdown of MDM2 in G/G cells moderately increased expression of subsets of p53 target genes without increasing p53 stability. Importantly, etoposide and actinomycin D treatments increased histone H3K36 trimethylation in T/T, but not G/G cells, suggesting a G/G correlated inhibition of transcription elongation. We therefore tested a chemotherapeutic agent (8-amino-adenosine) that induces p53-independent cell death for higher clinically relevant cytotoxicity. We demonstrated that T/T and G/G mdm2 SNP309 cells were equally sensitive to 8-amino-adenosine induced cell death. In conclusion for cancer cells overexpressing MDM2, targeting MDM2 may be less effective than inducing p53-independent cell death.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, Molecular Biology, suitable for electrophoresis
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture