Merck
  • Bio-sample detection on paper-based devices with inkjet printer-sprayed reagents.
Bio-sample detection on paper-based devices with inkjet printer-sprayed reagents.
Talanta (2015-10-16)
Wun-Hong Liang, Chien-Hung Chu, Ruey-Jen Yang
ABSTRACT
The reagent required for bio-sample detection on paper-based analytical devices is generally introduced manually using a pipette. Such an approach is time-consuming; particularly if a large number of devices are required. Automated methods provide a far more convenient solution for large-scale production, but incur a substantial cost. Accordingly, the present study proposes a low-cost method for the paper-based analytical devices in which the biochemical reagents are sprayed onto the device directly using a modified commercial inkjet printer. The feasibility of the proposed method is demonstrated by performing aspartate aminotransferase (AST) and alanine aminotransferase (ALT) tests using simple two-dimensional (2D) paper-based devices. In both cases, the reaction process is analyzed using an image-processing-based colorimetric method. The experimental results show that for AST detection within the 0-105 U/l concentration range, the optimal observation time is around four minutes, while for ALT detection in the 0-125 U/l concentration range, the optimal observation time is approximately one minute. Finally, for both samples, the detection performance of the sprayed-reagent analytical devices is insensitive to the glucose concentration.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Methyl Green, zinc chloride salt, certified by the BSC, Dye content 85 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, <0.5% crystal violet, Dye content 80 %
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Rhodamine B, suitable for fluorescence
Sigma-Aldrich
Methyl Green, zinc chloride salt, suitable for microscopy
Sigma-Aldrich
4-Aminoantipyrine, puriss. p.a., reag. Ph. Eur., ≥99%
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
4-Aminoantipyrine, reagent grade
Sigma-Aldrich
Methyl Green, zinc chloride salt, ~85%
Sigma-Aldrich
Rhodamine B, Dye content 90 %
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Thiamine pyrophosphate, ≥95%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%